Assay Details
| Gene Symbol: Mbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010776.1 | NP_034906.1 | 3 - 4 | 348 | 81 | |
| GenBank mRNA | AK149484.1 | - | 3 - 4 | 378 | 81 | |
| BC010760.1 | - | 3 - 4 | 363 | 4212216 | 81 | |
| D11440.1 | - | 3 - 4 | 348 | 81 | ||
| S42294.1 | - | 3 - 4 | 374 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbl2
Genomic Map