Assay Details
| Gene Symbol: Gm13089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC053717.1 | - | 2 - 3 | 378 | 131 | |
| BC094933.1 | - | 2 - 3 | 370 | 131 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988093 | BC094933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988093 | BC094933.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03541745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03396845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03396855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03401465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03932431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03541744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03541743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03396841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03647885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03396846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03401469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03647887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03510323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03647886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03396858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23375944 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 359749 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258764 | - | - |

Gm13089
Genomic Map