Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 10 - 11 | 1399 | 60 | |
| GenBank mRNA | AK082884.1 | - | 10 - 11 | 1390 | 60 | |
| BC027065.1 | - | 5 - 6 | 473 | 60 | ||
| BC068126.1 | - | 10 - 11 | 1307 | 6829456 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tyw1
Genomic Map