Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 1 - 2 | 162 | 75 | |
| GenBank mRNA | AK082884.1 | - | 1 - 2 | 153 | 75 | |
| AK162899.1 | - | 1 - 2 | 165 | 75 | ||
| BB619468.1 | - | 1 - 2 | 162 | 75 | ||
| BC068126.1 | - | 1 - 2 | 70 | 6829456 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082884.1, AK162899.1, BC068126.1, NM_001015876.2 transcripts.
Set Membership: |
|

Tyw1
Genomic Map