Assay Details
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145218.3 | NP_660253.2 | 7 - 8 | 2193 | 140 | |
GenBank mRNA | AB175684.1 | - | 7 - 8 | 1271 | 140 | |
AF467979.1 | - | 7 - 8 | 1474 | 140 | ||
BC052469.1 | - | 7 - 8 | 1589 | 6810287 | 140 | |
BC069177.1 | - | 7 - 8 | 2193 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
