Assay Details
Gene Symbol: Zhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199449.2 | NP_955520.1 | 1 - 2 | 339 | 155 | |
GenBank mRNA | AK031782.1 | - | 1 - 2 | 236 | 155 | |
AK157631.1 | - | 1 - 2 | 326 | 155 | ||
BC059178.1 | - | 1 - 2 | 93 | 6400319 | 155 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK031782.1, AK157631.1, BC059178.1, NM_199449.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
