Assay Details
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 18 - 19 | 2064 | 87 | |
GenBank mRNA | BC060184.1 | - | 17 - 18 | 2505 | 87 | |
BC139010.1 | - | 18 - 19 | 1994 | 8862032 | 87 | |
BC139011.1 | - | 18 - 19 | 1994 | 8862033 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
