Assay Details
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 21 - 22 | 2426 | 95 | |
GenBank mRNA | AJ005678.1 | - | 21 - 22 | 2384 | 95 | |
AK145898.1 | - | 18 - 19 | 2660 | 95 | ||
BC060184.1 | - | 20 - 21 | 2867 | 95 | ||
BC139010.1 | - | 21 - 22 | 2356 | 8862032 | 95 | |
BC139011.1 | - | 21 - 22 | 2356 | 8862033 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
