Assay Details
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 29 - 30 | 3440 | 125 | |
GenBank mRNA | AJ005678.1 | - | 29 - 30 | 3398 | 125 | |
AK132652.1 | - | 4 - 5 | 495 | 125 | ||
AK145898.1 | - | 26 - 27 | 3674 | 125 | ||
BC060184.1 | - | 28 - 29 | 3881 | 125 | ||
BC139010.1 | - | 29 - 30 | 3370 | 8862032 | 125 | |
BC139011.1 | - | 29 - 30 | 3370 | 8862033 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
