Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500015173.mm.1 |
TC0500001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015173.mm.1 |
NM_016964.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015173.mm.1 |
NM_016964.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015180.mm.1 |
TC0500001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015180.mm.1 |
NM_016964.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015180.mm.1 |
NM_016964.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015177.mm.1 |
TC0500001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015177.mm.1 |
NM_016964.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015177.mm.1 |
NM_016964.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015178.mm.1 |
TC0500001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015178.mm.1 |
NM_016964.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015178.mm.1 |
NM_016964.2 |
Refseq |