Assay Details
| Gene Symbol: Gtf2ird2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053266.1 | NP_444496.1 | 14 - 15 | 1436 | 77 | |
| GenBank mRNA | AY014963.1 | - | 14 - 15 | 1436 | 77 | |
| AY116023.1 | - | 14 - 15 | 1447 | 77 | ||
| BC057179.1 | - | 13 - 14 | 1201 | 77 | ||
| BC132157.1 | - | 14 - 15 | 1344 | 40130434 | 77 | |
| BC132159.1 | - | 14 - 15 | 1344 | 40130436 | 77 | |
More Information
Set Membership: |
|

Gtf2ird2
Genomic Map