Assay Details
| Gene Symbol: Gtf2ird2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053266.1 | NP_444496.1 | 9 - 10 | 968 | 110 | |
| GenBank mRNA | AK082488.1 | - | 1137 | 110 | ||
| AY014963.1 | - | 9 - 10 | 968 | 110 | ||
| AY116023.1 | - | 9 - 10 | 979 | 110 | ||
| BC132157.1 | - | 9 - 10 | 876 | 40130434 | 110 | |
| BC132159.1 | - | 9 - 10 | 876 | 40130436 | 110 | |
More Information
Set Membership: |
|

Gtf2ird2
Genomic Map