Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800004057.mm.1 |
NM_001005847.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004057.mm.1 |
NM_001005847.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004057.mm.1 |
NM_001205054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004057.mm.1 |
NM_001205054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004057.mm.1 |
TC0800000585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800004058.mm.1 |
NM_001005847.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004058.mm.1 |
NM_001005847.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004058.mm.1 |
NM_001205054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004058.mm.1 |
NM_001205054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004058.mm.1 |
TC0800000585.mm.1 |
MTA TC |