Assay Details
| Gene Symbol: Cnot6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285511.1 | NP_001272440.1 | 7 - 8 | 894 | 84 | |
| NM_001285514.1 | NP_001272443.1 | 7 - 8 | 768 | 84 | ||
| NM_144910.2 | NP_659159.1 | 7 - 8 | 762 | 84 | ||
| NM_178854.4 | NP_849185.2 | 7 - 8 | 772 | 84 | ||
| GenBank mRNA | AK030112.1 | - | 7 - 8 | 770 | 84 | |
| AK207400.1 | - | 3 - 4 | 299 | 84 | ||
| BC018506.1 | - | 7 - 8 | 762 | 3598248 | 84 | |
| BC023923.1 | - | 3 - 4 | 284 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnot6l
Genomic Map