Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500030779.mm.1 |
TC0500003406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030779.mm.1 |
NM_001289554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030779.mm.1 |
NM_001289555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030779.mm.1 |
NM_029659.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030779.mm.1 |
NM_029659.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
TC0500003406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
NM_001289554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
NM_001289555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
NM_001289556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
NM_001289557.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
NM_029659.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030780.mm.1 |
NM_029659.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
TC0500003406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
NM_001289554.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
NM_001289555.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
NM_001289556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
NM_001289557.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
NM_029659.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030787.mm.1 |
NM_029659.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030789.mm.1 |
TC0500003406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030789.mm.1 |
NM_001289556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030789.mm.1 |
NM_001289557.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030789.mm.1 |
NM_029659.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030789.mm.1 |
NM_029659.4 |
Refseq |