Assay Details
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027891.4 | NP_082167.2 | 7 - 8 | 1043 | 65 | |
GenBank mRNA | AK004717.1 | - | 5 - 6 | 2005 | 65 | |
AK051910.1 | - | 7 - 8 | 991 | 65 | ||
AK084948.1 | - | 7 - 8 | 1043 | 65 | ||
AK153976.1 | - | 7 - 8 | 1014 | 65 | ||
AK154352.1 | - | 7 - 8 | 985 | 65 | ||
AK167020.1 | - | 7 - 8 | 992 | 65 | ||
AK172708.1 | - | 7 - 8 | 1011 | 65 | ||
BC048397.1 | - | 7 - 8 | 1022 | 6396815 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
