Assay Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008617.2 | NP_032643.2 | 4 - 5 | 520 | 77 | |
| GenBank mRNA | AK002305.1 | - | 4 - 5 | 537 | 77 | |
| AK135162.1 | - | 4 - 5 | 533 | 77 | ||
| AK160553.1 | - | 4 - 5 | 503 | 77 | ||
| AK167809.1 | - | 4 - 5 | 503 | 77 | ||
| AK204746.1 | - | 2 - 3 | 198 | 77 | ||
| BC023482.1 | - | 4 - 5 | 520 | 5251192 | 77 | |
| DQ402950.1 | - | 3 - 4 | 370 | 77 | ||
| M16229.1 | - | 4 - 5 | 479 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdh2
Genomic Map