Assay Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008617.2 | NP_032643.2 | 6 - 7 | 725 | 76 | |
| GenBank mRNA | AK135162.1 | - | 6 - 7 | 738 | 76 | |
| AK160553.1 | - | 6 - 7 | 708 | 76 | ||
| AK167809.1 | - | 6 - 7 | 708 | 76 | ||
| BC023482.1 | - | 6 - 7 | 725 | 5251192 | 76 | |
| DQ402950.1 | - | 5 - 6 | 575 | 76 | ||
| M16229.1 | - | 6 - 7 | 684 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdh2
Genomic Map