Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148932.2 | NP_683734.2 | 5 - 6 | 1235 | 68 | |
| GenBank mRNA | AF516680.1 | - | 5 - 6 | 1199 | 68 | |
| AK154473.1 | - | 5 - 6 | 1236 | 68 | ||
| AK172982.1 | - | 4 - 5 | 613 | 68 | ||
| BC053101.1 | - | 5 - 6 | 1211 | 6405486 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pom121
Genomic Map