Assay Details
Gene Symbol: Slc15a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023044.2 | NP_075531.2 | 3 - 4 | 1468 | 64 | |
GenBank mRNA | AF121080.1 | - | 3 - 4 | 1476 | 64 | |
AK051941.1 | - | 3 - 4 | 1069 | 64 | ||
AK171698.1 | - | 1121 | 64 | |||
BC051940.1 | - | 3 - 4 | 1139 | 5717926 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
