Assay Details
Gene Symbol: Slc15a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023044.2 | NP_075531.2 | 7 - 8 | 2060 | 87 | |
GenBank mRNA | AF121080.1 | - | 7 - 8 | 2068 | 87 | |
AK051941.1 | - | 7 - 8 | 1661 | 87 | ||
BC051940.1 | - | 7 - 8 | 1731 | 5717926 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | AK051941.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | BC051940.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | NM_023044.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | AF121080.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | AK051941.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | BC051940.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | NM_023044.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P467389 | AF121080.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89754 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295216 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292880 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877546 | AK049964.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357707 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539711 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946243 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243316 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745867 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148103 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175123 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135590 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | AK051941.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | BC051940.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | NM_023044.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | AF121080.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | AK051941.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | BC051940.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | NM_023044.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P467389 | AF121080.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953154 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953180 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09105023 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175123 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135590 | - | - |
Whole Mouse Genome G4122A | A_52_P467389 | AK051941.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | BC051940.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | NM_023044.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | AF121080.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | AK051941.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | BC051940.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | NM_023044.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P467389 | AF121080.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108951 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124524 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175123 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135590 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378165 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378172 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378155 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378166 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378169 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378156 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378174 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14378170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14385240 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953161 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14196717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953227 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14256404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14446656 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14213082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14164502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14164496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14419285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14243494 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14378174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31953203 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10461559 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461564 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461565 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461561 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461567 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461560 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461566 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461562 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461563 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420697_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10461558 | - | - |
Mouse Genome 430 2.0 Array | 1420697_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420697_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65221_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65221_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900002653.mm.1 | NM_023044.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002653.mm.1 | NM_023044.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002653.mm.1 | TC1900000211.mm.1 | MTA TC |
Mouse Expression Set 430 | 1420697_PM_at | - | - |
Mouse Expression Set 430 | 1420697_at | - | - |
Mouse Genome 430A 2.0 Array | 1420697_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420697_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420697_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420697_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 420356 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6650333 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6650333 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006650333 | - | - |
mouse-6 v1.1 (Illumina) | scl53366.9.1_53-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6650333 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl53366.9.1_53-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650333 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2987709 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2987709 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707798 | - | - |
