Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900002546.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002546.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002546.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002546.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002546.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002552.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002552.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002552.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002552.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002545.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002545.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002545.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002545.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002545.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002543.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002543.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002543.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002543.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002543.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002555.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002555.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002555.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002555.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002555.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002549.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002549.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002549.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002549.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002549.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002550.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002550.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002550.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002550.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002547.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002547.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002547.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002547.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002553.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002553.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002553.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002553.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002553.mm.1 |
TC1900000204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900002554.mm.1 |
NM_001282064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002554.mm.1 |
NM_001282065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002554.mm.1 |
NM_001282067.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002554.mm.1 |
NM_201351.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900002554.mm.1 |
TC1900000204.mm.1 |
MTA TC |