Assay Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282064.1 | NP_001268993.1 | 5 - 6 | 1339 | 65 | |
| NM_001282065.1 | NP_001268994.1 | 4 - 5 | 1436 | 65 | ||
| NM_001282067.1 | NP_001268996.1 | 6 - 7 | 1359 | 65 | ||
| NM_201351.2 | NP_958739.1 | 5 - 6 | 1529 | 65 | ||
| GenBank mRNA | AK040692.1 | - | 1233 | 65 | ||
| AK139533.1 | - | 5 - 6 | 1517 | 65 | ||
| AK160179.1 | - | 4 - 5 | 1140 | 65 | ||
| AK168841.1 | - | 5 - 6 | 1233 | 65 | ||
| AK169729.1 | - | 5 - 6 | 1235 | 65 | ||
| AK170911.1 | - | 6 - 7 | 1027 | 65 | ||
| AK172638.1 | - | 5 - 6 | 1046 | 65 | ||
| BC065078.1 | - | 5 - 6 | 963 | 6817093 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb561a3
Genomic Map