Assay Details
Gene Symbol: Cyb561a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282064.1 | NP_001268993.1 | 6 - 7 | 1497 | 93 | |
NM_001282065.1 | NP_001268994.1 | 5 - 6 | 1594 | 93 | ||
NM_001282067.1 | NP_001268996.1 | 7 - 8 | 1517 | 93 | ||
NM_201351.2 | NP_958739.1 | 6 - 7 | 1687 | 93 | ||
GenBank mRNA | AK040692.1 | - | 1391 | 93 | ||
AK139533.1 | - | 6 - 7 | 1675 | 93 | ||
AK160179.1 | - | 5 - 6 | 1298 | 93 | ||
AK168841.1 | - | 6 - 7 | 1391 | 93 | ||
AK169729.1 | - | 6 - 7 | 1393 | 93 | ||
AK170911.1 | - | 7 - 8 | 1185 | 93 | ||
AK172638.1 | - | 6 - 7 | 1204 | 93 | ||
BC065078.1 | - | 6 - 7 | 1121 | 6817093 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
biological_process
oxidation-reduction process
molecular_function
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
metal ion binding
epoxyqueuosine reductase activity
transport
biological_process
oxidation-reduction process
molecular_function
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
metal ion binding
epoxyqueuosine reductase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | BC065078.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK170911.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK172638.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK168841.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | BC065078.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK170911.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK172638.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK168841.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011355 | AK040692.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354138 | AK019697.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652443 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10120 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355598 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143178 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123376 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | BC065078.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK170911.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK172638.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK168841.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK169729.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | BC065078.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK170911.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK172638.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK168841.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | BC065078.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK170911.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK172638.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK168841.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | BC065078.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK170911.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK172638.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK168841.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951480 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951495 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951488 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951466 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951485 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09104225 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09104215 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03549246 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123376 | - | - |
Whole Mouse Genome G4122A | A_51_P324871 | BC065078.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK170911.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK172638.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK168841.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | BC065078.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK170911.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK172638.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK168841.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P324871 | AK169729.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115859 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123376 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14355982 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14355984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951485 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951494 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951498 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14355984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951486 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951480 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951465 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14316645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31951479 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10461488 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461492 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461495 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461489 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461491 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461496 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461493 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461490 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10461494 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454895_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434245_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447244_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10461487 | - | - |
Mouse Genome 430 2.0 Array | 1454895_at | - | - |
Mouse Genome 430 2.0 Array | 1434245_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1434245_a_at | - | - |
Mouse Genome 430 2.0 Array | 1447244_at | - | - |
Mouse Genome 430 2.0 Array | 1454895_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447244_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225912_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225912_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900002559.mm.1 | NM_001282064.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002559.mm.1 | NM_001282067.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002559.mm.1 | NM_201351.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002559.mm.1 | TC1900000204.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900002557.mm.1 | NM_001282064.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002557.mm.1 | NM_001282065.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002557.mm.1 | NM_001282067.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002557.mm.1 | NM_201351.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002557.mm.1 | TC1900000204.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900002560.mm.1 | NM_001282064.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002560.mm.1 | NM_001282067.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002560.mm.1 | NM_201351.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900002560.mm.1 | TC1900000204.mm.1 | MTA TC |
Mouse Expression Set 430 | 1454895_PM_at | - | - |
Mouse Expression Set 430 | 1434245_PM_a_at | - | - |
Mouse Expression Set 430 | 1454895_at | - | - |
Mouse Expression Set 430 | 1447244_at | - | - |
Mouse Expression Set 430 | 1447244_PM_at | - | - |
Mouse Expression Set 430 | 1434245_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447244_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454895_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454895_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447244_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434245_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434245_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5420110 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005420110 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2590780 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2590780 | - | - |
mouse-6 v1.1 (Illumina) | scl0225912.8_241-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5420110 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0225912.8_241-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420110 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2590779 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2590779 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0225912.8_241-S | - | - |
