Assay Details
| Gene Symbol: Cds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173370.3 | NP_775546.2 | 10 - 11 | 1276 | 79 | |
| GenBank mRNA | AF533367.2 | - | 10 - 11 | 1185 | 79 | |
| BC026505.1 | - | 5 - 6 | 443 | 79 | ||
| BC055292.1 | - | 10 - 11 | 1276 | 6491596 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cds1
Genomic Map