Assay Details
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009177.4 | NP_033203.1 | 6 - 7 | 1666 | 91 | |
GenBank mRNA | AK031344.1 | - | 6 - 7 | 1556 | 91 | |
AK078469.1 | - | 5 - 6 | 975 | 91 | ||
AK138747.1 | - | 6 - 7 | 1615 | 91 | ||
BC023130.1 | - | 5 - 6 | 1918 | 91 | ||
BC040763.1 | - | 3 - 4 | 719 | 91 | ||
BC084730.1 | - | 3 - 4 | 867 | 30355523 | 91 | |
BC099670.1 | - | 6 - 7 | 1553 | 4187038 | 91 | |
BC099693.1 | - | 7 - 8 | 1633 | 4910140 | 91 | |
X73523.1 | - | 3 - 4 | 684 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
