Assay Details
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009177.4 | NP_033203.1 | 7 - 8 | 1700 | 87 | |
GenBank mRNA | AK031344.1 | - | 7 - 8 | 1590 | 87 | |
AK078469.1 | - | 6 - 7 | 1009 | 87 | ||
AK138747.1 | - | 7 - 8 | 1649 | 87 | ||
BC023130.1 | - | 6 - 7 | 1952 | 87 | ||
BC040763.1 | - | 4 - 5 | 753 | 87 | ||
BC084730.1 | - | 4 - 5 | 901 | 30355523 | 87 | |
BC099670.1 | - | 7 - 8 | 1587 | 4187038 | 87 | |
BC099693.1 | - | 8 - 9 | 1667 | 4910140 | 87 | |
X73523.1 | - | 4 - 5 | 718 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
