Assay Details
Gene Symbol: 1700012I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045140.1 | 2 - 3 | 392 | 108 | ||
GenBank mRNA | AK005914.1 | - | 2 - 3 | 393 | 108 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY706053.1 | - | 396 | Chr. - - [-] on Build | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ST3 beta-galactoside alpha-2,3-sialyltransferase 1 | St3gal1 |
5330418N22Rik, AI467004, ST3GalI, Siat4, Siat4a, St3gal-1 |
-62.7kb | Anti-sense | Chr. 15 - 67102875 - 67176910 on NCBI Build GRCm38 |
Downstream | zinc finger and AT hook domain containing | Zfat |
Gm922, Zfat1, Zfp406 |
832.5kb | Anti-sense | Chr. 15 - 68083714 - 68259265 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
