Assay Details
Gene Symbol: 1700012I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045140.1 | 8 - 9 | 856 | 93 | ||
GenBank mRNA | AK005914.1 | - | 8 - 9 | 856 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ST3 beta-galactoside alpha-2,3-sialyltransferase 1 | St3gal1 |
5330418N22Rik, AI467004, ST3GalI, Siat4, Siat4a, St3gal-1 |
-196.0kb | Anti-sense | Chr. 15 - 67102875 - 67176910 on NCBI Build GRCm38 |
Downstream | zinc finger and AT hook domain containing | Zfat |
Gm922, Zfat1, Zfp406 |
706.6kb | Anti-sense | Chr. 15 - 68083714 - 68259265 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095380 | AV040489.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095380 | AV208344.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112849 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499842 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223137 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240746 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445575 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972926 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910261 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381423 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805805 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652681 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876007 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956755 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158782 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17667 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80225 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718150 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285064 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303813 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924081 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357317 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676355 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518480 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375195 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415167 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933536 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939047 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13184 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740788 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45490 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497704 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311953 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525449 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280404 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457692 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89946 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149139 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849563 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224972 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78195 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235684 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805638 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62441 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77730 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154572 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11139 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 709 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33155 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159147 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136761 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095380 | AV040489.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095380 | AV208344.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08401518 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136761 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03058919 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03058982 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08401225 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08401492 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03058947 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08401444 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03058966 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03058881 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08401316 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03058982 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03058919 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136761 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03058947 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03058881 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1453460_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453460_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453460_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500003528.mm.1 | NR_045140.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003528.mm.1 | NR_045140.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003528.mm.1 | TC1500000496.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500003527.mm.1 | NR_045140.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003527.mm.1 | NR_045140.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500003527.mm.1 | TC1500000496.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453460_PM_at | - | - |
Mouse Expression Set 430 | 1453460_at | - | - |
Murine Genome U74v2 Set | 168582_f_at | - | - |
Murine Genome U74v2 Set | 169309_PM_i_at | - | - |
Murine Genome U74v2 Set | 169309_i_at | - | - |
Murine Genome U74v2 Set | 168582_PM_f_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453460_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453460_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104730008 | - | - |
