Assay Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172711.2 | NP_766299.1 | 7 - 8 | 799 | 67 | |
| GenBank mRNA | AK032597.1 | - | 6 - 7 | 1807 | 67 | |
| AK047411.1 | - | 7 - 8 | 668 | 67 | ||
| AK053420.1 | - | 7 - 8 | 773 | 67 | ||
| AK140534.1 | - | 7 - 8 | 758 | 67 | ||
| AK160956.1 | - | 7 - 8 | 818 | 67 | ||
| AK165012.1 | - | 7 - 8 | 788 | 67 | ||
| BC057962.1 | - | 7 - 8 | 753 | 5353891 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Guf1
Genomic Map