Assay Details
| Gene Symbol: Ldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039072.2 | NP_001034161.1 | 6 - 7 | 653 | 99 | |
| GenBank mRNA | AY206013.1 | - | 6 - 7 | 617 | 99 | |
More Information
Set Membership: |
|

Ldb3
Genomic Map