Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 8 - 9 | 873 | 108 | |
| GenBank mRNA | AF263460.1 | - | 6 - 7 | 427 | 108 | |
| AK028885.1 | - | 8 - 9 | 840 | 108 | ||
| AK035541.1 | - | 8 - 9 | 860 | 108 | ||
| AK082853.1 | - | 8 - 9 | 875 | 108 | ||
| AK143277.1 | - | 8 - 9 | 874 | 108 | ||
| AK149549.1 | - | 1 - 2 | 1024 | 108 | ||
| AY682914.1 | - | 9 - 10 | 866 | 108 | ||
| BC026565.1 | - | 4 - 5 | 291 | 108 | ||
| BC027806.1 | - | 8 - 9 | 635 | 108 | ||
| BC031705.1 | - | 5 - 6 | 387 | 108 | ||
| BC055773.1 | - | 5 - 6 | 326 | 108 | ||
| BC078440.1 | - | 6 - 7 | 450 | 108 | ||
| BC141008.1 | - | 9 - 10 | 839 | 9055620 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a9
Genomic Map