Assay Details
| Gene Symbol: Nsun7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014968.1 | - | 10 - 11 | 1314 | 138 | |
| AK035181.1 | - | 10 - 11 | 1727 | 138 | ||
| BC115878.1 | - | 10 - 11 | 1638 | 40088737 | 138 | |
| BC116658.1 | - | 10 - 11 | 1638 | 40088738 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nsun7
Genomic Map