Assay Details
| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009466.2 | NP_033492.1 | 3 - 4 | 494 | 72 | |
| GenBank mRNA | AF061017.1 | - | 3 - 4 | 378 | 72 | |
| AK146693.1 | - | 3 - 4 | 378 | 72 | ||
| AK162298.1 | - | 3 - 4 | 382 | 72 | ||
| AK167462.1 | - | 3 - 4 | 374 | 72 | ||
| AK167560.1 | - | 3 - 4 | 474 | 72 | ||
| AK168064.1 | - | 1 - 2 | 91 | 72 | ||
| AK168508.1 | - | 3 - 4 | 381 | 72 | ||
| AK168563.1 | - | 3 - 4 | 378 | 72 | ||
| AK169208.1 | - | 3 - 4 | 509 | 72 | ||
| BC006749.1 | - | 3 - 4 | 494 | 3708709 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugdh
Genomic Map