Assay Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161420.1 | NP_001154892.1 | 11 - 12 | 2293 | 65 | |
| NM_013703.2 | NP_038731.2 | 11 - 12 | 2293 | 65 | ||
| GenBank mRNA | AK044683.1 | - | 3 - 4 | 901 | 65 | |
| AK076172.1 | - | 11 - 12 | 2298 | 65 | ||
| AK136838.1 | - | 11 - 12 | 2113 | 65 | ||
| BC013622.1 | - | 11 - 12 | 1729 | 3968213 | 65 | |
| L33417.1 | - | 11 - 12 | 1717 | 65 | ||
| U06670.1 | - | 11 - 12 | 1879 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vldlr
Genomic Map