Assay Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161420.1 | NP_001154892.1 | 9 - 10 | 1906 | 101 | |
| NM_013703.2 | NP_038731.2 | 9 - 10 | 1906 | 101 | ||
| GenBank mRNA | AK076172.1 | - | 9 - 10 | 1911 | 101 | |
| AK136838.1 | - | 9 - 10 | 1726 | 101 | ||
| BC013622.1 | - | 9 - 10 | 1342 | 3968213 | 101 | |
| L33417.1 | - | 9 - 10 | 1330 | 101 | ||
| U06670.1 | - | 9 - 10 | 1492 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vldlr
Genomic Map