Assay Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281969.1 | NP_001268898.1 | 6 - 7 | 725 | 80 | |
| NM_010298.6 | NP_034428.2 | 6 - 7 | 775 | 80 | ||
| GenBank mRNA | AK082578.1 | - | 6 - 7 | 778 | 80 | |
| AK083251.1 | - | 5 - 6 | 629 | 80 | ||
| AM420316.1 | - | 5 - 6 | 608 | 80 | ||
| BC027094.1 | - | 6 - 7 | 729 | 80 | ||
| BC037605.1 | - | 6 - 7 | 743 | 5361798 | 80 | |
| U09399.1 | - | 6 - 7 | 684 | 80 | ||
| X81202.1 | - | 5 - 6 | 567 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glrb
Genomic Map