Assay Details
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023134.4 | NP_075623.2 | 1 - 2 | 46 | 60 | |
GenBank mRNA | AK004620.1 | - | 1 - 2 | 47 | 60 | |
AK004788.1 | - | 1 - 2 | 47 | 60 | ||
BC061096.1 | - | 1 - 2 | 46 | 30300708 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK004620.1, AK004788.1, BC061096.1, NM_023134.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
