Assay Details
| Gene Symbol: Sftpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023134.4 | NP_075623.2 | 3 - 4 | 278 | 87 | |
| GenBank mRNA | AK004620.1 | - | 3 - 4 | 279 | 87 | |
| AK004788.1 | - | 3 - 4 | 279 | 87 | ||
| AK011333.1 | - | 2 - 3 | 252 | 87 | ||
More Information
Set Membership: |
|

Sftpa1
Genomic Map