Assay Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029884.1 | NP_084160.1 | 2 - 3 | 337 | 91 | |
| GenBank mRNA | AK035264.1 | - | 2 - 3 | 270 | 91 | |
| AK149883.1 | - | 2 - 3 | 341 | 91 | ||
| AK152926.1 | - | 2 - 3 | 302 | 91 | ||
| AK159649.1 | - | 2 - 3 | 266 | 91 | ||
| AK160068.1 | - | 2 - 3 | 260 | 91 | ||
| BC024084.1 | - | 2 - 3 | 244 | 5101064 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgsnat
Genomic Map