Assay Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029884.1 | NP_084160.1 | 3 - 4 | 474 | 99 | |
| GenBank mRNA | AK035264.1 | - | 3 - 4 | 407 | 99 | |
| AK149883.1 | - | 3 - 4 | 478 | 99 | ||
| AK152926.1 | - | 3 - 4 | 439 | 99 | ||
| AK159649.1 | - | 3 - 4 | 403 | 99 | ||
| AK160068.1 | - | 3 - 4 | 397 | 99 | ||
| AK198139.1 | - | 1 - 2 | 115 | 99 | ||
| BC024084.1 | - | 3 - 4 | 381 | 5101064 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgsnat
Genomic Map