Assay Details
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145482.3 | NP_663457.2 | 10 - 11 | 1478 | 74 | |
GenBank mRNA | AY037804.1 | - | 10 - 11 | 1478 | 74 | |
BC029101.1 | - | 10 - 11 | 1993 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
