Assay Details
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145482.3 | NP_663457.2 | 1 - 2 | 251 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_145482.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
