Assay Details
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY037804.1 | - | 4 - 5 | 581 | 127 | |
BC029101.1 | - | 4 - 5 | 478 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
