Assay Details
Gene Symbol: 1700024G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034037.1 | NP_001029209.1 | 2 - 3 | 237 | 74 | |
GenBank mRNA | AK006301.1 | - | 2 - 3 | 256 | 74 | |
AK007014.1 | - | 2 - 3 | 255 | 74 | ||
AK019799.1 | - | 2 - 3 | 244 | 74 | ||
BC107222.2 | - | 2 - 3 | 234 | 40051877 | 74 | |
BC107223.1 | - | 2 - 3 | 237 | 40051878 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | AK006301.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | AK007014.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | AK019799.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | BC107222.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | BC107223.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | AK006301.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | AK007014.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | AK019799.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | BC107222.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P625270 | BC107223.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949796 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526574 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162545 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36185 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | AK006301.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | AK007014.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | AK019799.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | BC107222.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | BC107223.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | AK006301.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | AK007014.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | AK019799.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | BC107222.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P625270 | BC107223.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517385 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517394 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517387 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517401 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517388 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517391 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29517398 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02827129 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08075322 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08075297 | - | - |
Whole Mouse Genome G4122A | A_52_P625270 | AK006301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | AK007014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | AK019799.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | BC107222.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | BC107223.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | AK006301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | AK007014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | AK019799.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | BC107222.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P625270 | BC107223.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02827129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29517397 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11780863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517356 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11813413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11439771 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11141531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517349 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517397 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517365 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517375 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11614598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11439782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11141530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11717334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11813409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517382 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517362 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517394 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11188215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11188201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11589351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11488135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517358 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517385 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11253168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517360 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517373 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29517354 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10418816 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10418814 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10418815 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10418812 | - | - |
Affymetrix Mouse Clariom S Array | PSR1400013196.mm.1 | NM_001034037.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400013196.mm.1 | NM_001034037.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400013196.mm.1 | TC1400001702.mm.1 | MTA TC |
Murine Genome U74v2 Set | 168254_PM_at | - | - |
Murine Genome U74v2 Set | 168254_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 746891 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5420204 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160151 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3160151 | - | - |
mouse-6 v1.1 (Illumina) | GI_77404226-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2788707 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2788707 | - | - |
