Assay Details
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139145.4 | NP_631884.1 | 2 - 3 | 228 | 67 | |
GenBank mRNA | AB066227.1 | - | 1 - 2 | 88 | 67 | |
AK052424.1 | - | 3 - 4 | 523 | 67 | ||
AK158212.1 | - | 3 - 4 | 345 | 67 | ||
AK164351.1 | - | 3 - 4 | 507 | 67 | ||
BC050090.1 | - | 2 - 3 | 229 | 30012868 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
