Assay Details
| Gene Symbol: Gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019521.2 | NP_062394.2 | 4 - 5 | 529 | 116 | |
| GenBank mRNA | AK086187.1 | - | 4 - 5 | 523 | 116 | |
| BC005444.1 | - | 4 - 5 | 484 | 3592398 | 116 | |
| X59846.1 | - | 4 - 5 | 500 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas6
Genomic Map