Assay Details
Gene Symbol: Gas6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019521.2 | NP_062394.2 | 5 - 6 | 657 | 90 | |
GenBank mRNA | AK086187.1 | - | 5 - 6 | 651 | 90 | |
BC005444.1 | - | 5 - 6 | 612 | 3592398 | 90 | |
X59846.1 | - | 5 - 6 | 628 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
