Assay Details
| Gene Symbol: Gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019521.2 | NP_062394.2 | 9 - 10 | 1142 | 65 | |
| GenBank mRNA | AK086187.1 | - | 9 - 10 | 1137 | 65 | |
| BC005444.1 | - | 9 - 10 | 1097 | 3592398 | 65 | |
| X59846.1 | - | 9 - 10 | 1113 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas6
Genomic Map