Assay Details
| Gene Symbol: Erc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177814.4 | NP_808482.2 | 9 - 10 | 2232 | 150 | |
| GenBank mRNA | AK122265.1 | - | 7 - 8 | 944 | 150 | |
| AK147213.1 | - | 10 - 11 | 2271 | 150 | ||
| AY356530.1 | - | 8 - 9 | 1919 | 150 | ||
| AY356531.1 | - | 8 - 9 | 1919 | 150 | ||
| AY356532.1 | - | 8 - 9 | 1919 | 150 | ||
| AY356533.1 | - | 8 - 9 | 1919 | 150 | ||
| BC056760.1 | - | 9 - 10 | 2232 | 6331997 | 150 | |
More Information
Set Membership: |
|

Erc2
Genomic Map